tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate 95%

Lieferant: Apollo Scientific

OR924245-1G
APOSOR924245-1GEA 487 EUR
APOSOR924245-1G
tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate 95%
tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954747
CAS-Nummer: 178152-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR