5-Bromo-2-vinylpyrimidine 95%
Lieferant: Apollo Scientific
OR924287-1G
APOSOR924287-1GEA
487
EUR
APOSOR924287-1G
5-Bromo-2-vinylpyrimidine 95%
5-Bromo-2-vinylpyrimidine
Formel:
C₆H₅BrN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20485168 CAS-Nummer: 883901-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...