(3-Bromothiophen-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR924308-1G OR924308-5G
APOSOR924308-1GEA 87 EUR
APOSOR924308-1G APOSOR924308-5G
(3-Bromothiophen-2-yl)methanol 95%
(3-Bromothiophen-2-yl)methanol
Formel: C₅H₅BrOS
Molecular Weight: 193.062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202660
CAS-Nummer: 70260-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR