(1-Propyl-1H-imidazol-5-yl)methanol 95%
Lieferant: Apollo Scientific
OR924312-1G
APOSOR924312-1GEA
574
EUR
APOSOR924312-1G
(1-Propyl-1H-imidazol-5-yl)methanol 95%
(1-Propyl-1H-imidazol-5-yl)methanol
Formel:
C₇H₁₂N₂O Molecular Weight: 140,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18808769 CAS-Nummer: 497855-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...