1-(2-Aminophenyl)pyrrol 95%

Lieferant: Apollo Scientific

Synonyme: 2-(1H-Pyrrol-1-yl)aniline

OR924329-5G
APOSOR924329-5GEA 62.5 EUR
APOSOR924329-5G
1-(2-Aminophenyl)pyrrol 95%
1-(2-Aminophenyl)pyrrole
Formel: C₁₀H₁₀N₂
Molecular Weight: 158.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005344
CAS-Nummer: 6025-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR