di-Natrium-β-glycerophosphat Hydrat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
ortho-Phosphorsäure(2-hydroxy-1-(hydroxymethyl)ethyl)ester Dinatriumsalz Hydrat
OR924358-25G
OR924358-100G
OR924358-500G
OR924358-250G
APOSOR924358-25GEA
132
EUR
APOSOR924358-25G
APOSOR924358-100G
APOSOR924358-500G
APOSOR924358-250G
di-Natrium-β-glycerophosphat Hydrat 98%
di-Natrium-β-glycerophosphat Hydrat
Phosphate group donor.
Formel:
C₃H₇Na₂O₆P·H₂O Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149082 CAS-Nummer: 154804-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...