di-Natrium-β-glycerophosphat Hydrat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: ortho-Phosphorsäure(2-hydroxy-1-(hydroxymethyl)ethyl)ester Dinatriumsalz Hydrat

OR924358-25G OR924358-100G OR924358-500G OR924358-250G
APOSOR924358-25GEA 132 EUR
APOSOR924358-25G APOSOR924358-100G APOSOR924358-500G APOSOR924358-250G
di-Natrium-β-glycerophosphat Hydrat 98%
di-Natrium-β-glycerophosphat Hydrat

Phosphate group donor.

Formel: C₃H₇Na₂O₆P·H₂O
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149082
CAS-Nummer: 154804-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR