3,3-Dimethoxypropiononitril 93%

Lieferant: Apollo Scientific

Synonyme: 3,3-Dimethoxypropanenitrile

OR924380-5G OR924380-1G OR924380-10G
APOSOR924380-5GEA 287 EUR
APOSOR924380-5G APOSOR924380-1G APOSOR924380-10G
3,3-Dimethoxypropiononitril 93%
3,3-Dimethoxypropiononitril
Formel: C₅H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00044797
CAS-Nummer: 57597-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR