1-Azaspiro[3.4]octan-2-one 95%

Lieferant: Apollo Scientific

OR924408-1G OR924408-250MG
APOSOR924408-1GEA 806 EUR
APOSOR924408-1G APOSOR924408-250MG
1-Azaspiro[3.4]octan-2-one 95%
1-Azaspiro[3.4]octan-2-one
Formel: C₇H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD20384799
CAS-Nummer: 178242-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR