(2R,4R)-N-Boc-4-Hydroxy-2-methylpyrrolidine 97%

Lieferant: Apollo Scientific

OR924457-1G
APOSOR924457-1GEA 503 EUR
APOSOR924457-1G
(2R,4R)-N-Boc-4-Hydroxy-2-methylpyrrolidine 97%
(2R,4R)-N-Boc-4-Hydroxy-2-methylpyrrolidine
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201,263 g/mol
Schmelzpunkt: 42…44 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11109735
CAS-Nummer: 114676-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR