1-Phenyl-1-butanol 95%

Lieferant: Apollo Scientific

OR924460-1G OR924460-25G OR924460-5G
APOSOR924460-1GEA 0 EUR
APOSOR924460-1G APOSOR924460-25G APOSOR924460-5G
1-Phenyl-1-butanol 95%
1-Phenyl-1-butanol
Formel: C₁₀H₁₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00066173
CAS-Nummer: 614-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR