4-(Methylthio)benzonitril 95%
Lieferant: Apollo Scientific
OR924462-25G
OR924462-5G
APOSOR924462-25GEA
169
EUR
APOSOR924462-25G
APOSOR924462-5G
4-(Methylthio)benzonitril 95%
4-(Methylthio)benzonitril
Formel:
C₈H₇NS Molecular Weight: 149.22 g/mol Siedepunkt: 272 °C Schmelzpunkt: 59…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010381 CAS-Nummer: 21382-98-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...