N-Formyl-Dl-Phenylalanin 95%

Lieferant: Apollo Scientific

Synonyme: 2-Formamido-3-phenylpropanoic acid

OR924498-25G OR924498-5G
APOSOR924498-25GEA 192 EUR
APOSOR924498-25G APOSOR924498-5G
N-Formyl-Dl-Phenylalanin 95%
N-Formyl-Dl-Phenylalanin
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063281
CAS-Nummer: 4289-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR