2,2'-Bis(2-oxazoline) 95%

Lieferant: Apollo Scientific

OR924511-5G
APOSOR924511-5GEA 163 EUR
APOSOR924511-5G
2,2'-Bis(2-oxazoline) 95%
2,2'-Bis(2-oxazoline)
Formel: C₆H₈N₂O₂
Molecular Weight: 140.14 g/mol
Schmelzpunkt: 213 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00059930
CAS-Nummer: 36697-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR