5-(2-Furyl)-1,3,4-oxadiazol-2-amine 95%
Lieferant: Apollo Scientific
OR924531-1G
APOSOR924531-1GEA
290
EUR
APOSOR924531-1G
5-(2-Furyl)-1,3,4-oxadiazol-2-amine 95%
5-(2-Furyl)-1,3,4-oxadiazol-2-amine
Formel:
C₆H₅N₃O₂ Molecular Weight: 151,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01710630 CAS-Nummer: 7659-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...