Methyl 5-chlorothiophene-3-carboxylate 95%

Lieferant: Apollo Scientific

OR924536-1G
APOSOR924536-1GEA 50.9 EUR
APOSOR924536-1G
Methyl 5-chlorothiophene-3-carboxylate 95%
Methyl 5-chlorothiophene-3-carboxylate
Formel: C₆H₅ClO₂S
Molecular Weight: 176,621 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20922775
CAS-Nummer: 36157-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR