Methyl 6-oxabicyclo[3.1.0]hexane-3-carboxylate 95%

Lieferant: Apollo Scientific

OR924547-1G
APOSOR924547-1GEA 400 EUR
APOSOR924547-1G
Methyl 6-oxabicyclo[3.1.0]hexane-3-carboxylate 95%
Methyl 6-oxabicyclo[3.1.0]hexane-3-carboxylate
Formel: C₇H₁₀O₃
Molecular Weight: 142,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20488708
CAS-Nummer: 119245-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR