(4-Methylcyclohexyl)methanol (cis/trans-Gemisch) 98%

Lieferant: Apollo Scientific

OR924567-1G
APOSOR924567-1GEA 78.3 EUR
APOSOR924567-1G
(4-Methylcyclohexyl)methanol (cis/trans-Gemisch) 98%
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture)
Formel: C₈H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00152628
CAS-Nummer: 34885-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR