(E)-3-Bromoacrylic acid 95%

Lieferant: Apollo Scientific

OR924573-1G
APOSOR924573-1GEA 183 EUR
APOSOR924573-1G
(E)-3-Bromoacrylic acid 95%
(E)-3-Bromoacrylic acid
Formel: C₃H₃BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00218760
CAS-Nummer: 6213-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR