Acetanilid 95%

Lieferant: Apollo Scientific

Synonyme: N-Phenylacetamid, Ethansäurephenylamid

OR924603-100G
APOSOR924603-100GEA 125 EUR
APOSOR924603-100G
Acetanilid 95%
Acetanilid
Formel: C₈H₉NO
Molecular Weight: 135,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008674
CAS-Nummer: 103-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR