Methyl-4-(2-hydroxyethoxy)benzoat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-(2-hydroxyethoxy)benzoate, 4-(2-Hydroxyethoxy)benzoic Acid Methyl Ester

OR924639-1G
APOSOR924639-1GEA 78.3 EUR
APOSOR924639-1G
Methyl-4-(2-hydroxyethoxy)benzoat 95%
Methyl-4-(2-hydroxyethoxy)benzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075872
CAS-Nummer: 3204-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR