2,4,6-Trichlor-1,3,5-triazin 95%

Lieferant: Apollo Scientific

OR924668-100G
APOSOR924668-100GEA 0 EUR
APOSOR924668-100G
2,4,6-Trichlor-1,3,5-triazin 95%
2,4,6-Trichloro-1,3,5-triazine
Formel: C₃Cl₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00006046
CAS-Nummer: 108-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR