Scyllo inositol 99%

Lieferant: Apollo Scientific
Achtung

OR924672-250MG
APOSOR924672-250MGEA 173 EUR
APOSOR924672-250MG
Scyllo inositol 99%
Scyllo inositol
Formel: C₆H₁₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00065455
CAS-Nummer: 488-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR