2-Chloro-5-ethoxypyrimidine 95%
Lieferant: Apollo Scientific
OR924684-1G
APOSOR924684-1GEA
151
EUR
APOSOR924684-1G
2-Chloro-5-ethoxypyrimidine 95%
2-Chloro-5-ethoxypyrimidine
Formel:
C₆H₇ClN₂O Molecular Weight: 158.586 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697651 CAS-Nummer: 82153-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...