4-Methylcyclohexanamin (cis/trans-Gemisch)
Lieferant: Apollo Scientific
OR924724-1G
OR924724-25G
OR924724-5G
APOSOR924724-1GEA
29
EUR
APOSOR924724-1G
APOSOR924724-25G
APOSOR924724-5G
4-Methylcyclohexanamin (cis/trans-Gemisch)
4-Methylcyclohexylamine (cis- and trans- mixture)
Formel:
C₇H₁₅N Siedepunkt: 148…154 °C Dichte: 0,855 g/cm³ (25 °C) Flammpunkt: 27 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001495 CAS-Nummer: 6321-23-9 UN: 2734 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...