Ethyl-4-chlormethylbenzoat 95%
Lieferant: Apollo Scientific
OR924738-25G
APOSOR924738-25GEA
0
EUR
APOSOR924738-25G
Ethyl-4-chlormethylbenzoat 95%
Ethyl-4-(Chloromethyl)benzoate
Formel:
C₁₀H₁₁ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04337336 CAS-Nummer: 1201-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...