2-Boc-3,4-dihydro-1h-pyrrolo[1,2-a]pyrazine-6-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR924758-250MG OR924758-1G
APOSOR924758-250MGEA 319 EUR
APOSOR924758-250MG APOSOR924758-1G
2-Boc-3,4-dihydro-1h-pyrrolo[1,2-a]pyrazine-6-carboxylic acid 95%
2-Boc-3,4-dihydro-1h-pyrrolo[1,2-a]pyrazine-6-carboxylic acid
Formel: C₁₃H₁₈N₂O₄
MDL: MFCD22395729
CAS-Nummer: 1363380-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR