4-Ethyl-2-methoxyphenol 98%
Lieferant: Apollo Scientific
Synonyme:
1-Ethyl-4-hydroxy-3-methoxybenzol, 4-Ethylguajakol, 5-Ethyl-2-hydroxyanisol
OR924780-100G
APOSOR924780-100GEA
72.5
EUR
APOSOR924780-100G
4-Ethyl-2-methoxyphenol 98%
4-Ethyl-2-methoxyphenol
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038714 CAS-Nummer: 2785-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...