3-(1-Piperidinylmethyl)phenol 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Piperidin-1-ylmethyl)phenol
OR924807-25G
APOSOR924807-25GEA
173
EUR
APOSOR924807-25G
3-(1-Piperidinylmethyl)phenol 95%
3-(1-Piperidinylmethyl)phenol
Formel:
C₁₂H₁₇NO Molecular Weight: 191.27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800250 CAS-Nummer: 73279-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...