tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%

Lieferant: Apollo Scientific

OR924855-25G
APOSOR924855-25GEA 104 EUR
APOSOR924855-25G
tert-Butyl 3,5-dioxopiperidine-1-carboxylate 97%
tert-Butyl 3,5-dioxopiperidine-1-carboxylate
Formel: C₁₀H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD14706038
CAS-Nummer: 396731-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR