5-Norbornen-2,3-Dicarboxylsäure Monomethylester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl Hydrogen 5-Norbornene-2,3-dicarboxylate, 3-(Methoxycarbonyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid, 5-Norbornene-2,3-dicarboxylic Acid Monomethyl Ester

OR924889-5G
APOSOR924889-5GEA 288 EUR
APOSOR924889-5G
5-Norbornen-2,3-Dicarboxylsäure Monomethylester 95%
5-Norbornen-2,3-Dicarboxylsäure Monomethylester
Formel: C₁₀H₁₂O₄
Molecular Weight: 196.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00167743
CAS-Nummer: 36897-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR