N,N-Di-n-propylformamide 95%

Lieferant: Apollo Scientific

OR925014-25G
APOSOR925014-25GEA 134 EUR
APOSOR925014-25G
N,N-Di-n-propylformamide 95%
N,N-Di-n-propylformamide
Formel: HCON(CH₂CH₂CH₃)₂
MDL: MFCD00043518
CAS-Nummer: 6282-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR