5-(1-Piperidinyl)-1-pentanol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(5-Hydroxypentyl)piperidine

OR925020-1G OR925020-5G
APOSOR925020-1GEA 203 EUR
APOSOR925020-1G APOSOR925020-5G
5-(1-Piperidinyl)-1-pentanol 95%
1-Piperidinepentanol
Formel: C₁₀H₂₁NO
Molecular Weight: 171.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093640
CAS-Nummer: 2937-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR