2-Methyl-4-phenyl-2-butanol 99%

Lieferant: Apollo Scientific

Synonyme: 2-Phenylethyldimethylcarbinol, Dimethyl-beta-phenylethylcarbinol

OR925050-500G
APOSOR925050-500GEA 136 EUR
APOSOR925050-500G
2-Methyl-4-phenyl-2-butanol 99%
2-Methyl-4-phenyl-2-butanol
Formel: C₁₁H₁₆O
Molecular Weight: 164.25 g/mol
Schmelzpunkt: 30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004472
CAS-Nummer: 103-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR