N-Ethyl-N-Methylbenzylamin 96%
Lieferant: Apollo Scientific
Synonyme:
Benzyl(ethyl)methylamine
OR925064-25G
APOSOR925064-25GEA
183
EUR
APOSOR925064-25G
N-Ethyl-N-Methylbenzylamin 96%
N-Ethyl-N-Methylbenzylamin
Formel:
C₁₀H₁₅N Molecular Weight: 149,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051914 CAS-Nummer: 4788-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...