2,5-Dimethylterephthalonitril 95%

Lieferant: Apollo Scientific

Synonyme: 2,5-Dicyano-p-xylene

OR925067-5G
APOSOR925067-5GEA 221 EUR
APOSOR925067-5G
2,5-Dimethylterephthalonitril 95%
2,5-Dimethylterephthalonitrile
Formel: C₁₀H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00191411
CAS-Nummer: 39095-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR