cis-Jasmon 95%
Lieferant: Apollo Scientific
Synonyme:
(Z)-3-Methyl-2-(pent-2-en-1-yl)cyclopent-2-enone, cis-Jasmon
OR925069-100G
APOSOR925069-100GEA
169
EUR
APOSOR925069-100G
cis-Jasmon 95%
cis-Jasmon
Formel:
C₁₁H₁₆O Molecular Weight: 164,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001402 CAS-Nummer: 488-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...