3-(Methoxymethoxy)benzaldehyd 95%

Lieferant: Apollo Scientific

OR925108-5G
APOSOR925108-5GEA 364 EUR
APOSOR925108-5G
3-(Methoxymethoxy)benzaldehyd 95%
3-(Methoxymethoxy)benzaldehyde
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038425
CAS-Nummer: 13709-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR