3-Methoxybenzamid 95%
Lieferant: Apollo Scientific
Synonyme:
m-Anisamid, 3-Methoxybenzoesäureamid
OR925166-25G
APOSOR925166-25GEA
79.9
EUR
APOSOR925166-25G
3-Methoxybenzamid 95%
3-Methoxybenzamid
Formel:
C₈H₉NO₂ Molecular Weight: 151.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007986 CAS-Nummer: 5813-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...