3,4-Dichlorpyridazin
Lieferant: Apollo Scientific
OR925203-1G
APOSOR925203-1GEA
807
EUR
APOSOR925203-1G
3,4-Dichlorpyridazin
3,4-Dichloropyridazine
Formel:
C₄H₂Cl₂N₂ Molecular Weight: 148.978 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20040517 CAS-Nummer: 1677-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...