trans,trans-Muconsäure (trans, trans-2,4-Hexadiendisäure) 95%
Lieferant: Apollo Scientific
OR925263-10G
OR925263-1G
OR925263-5G
APOSOR925263-10GEA
0
EUR
APOSOR925263-10G
APOSOR925263-1G
APOSOR925263-5G
trans,trans-Muconsäure (trans, trans-2,4-Hexadiendisäure) 95%
trans,trans-Muconsäure (trans, trans-2,4-Hexadiendisäure)
Formel:
C₆H₆O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002702 CAS-Nummer: 3588-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...