(R)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
Lieferant: Apollo Scientific
OR925276-1G
APOSOR925276-1GEA
232
EUR
APOSOR925276-1G
(R)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
(R)-1-(3,4-Dimethoxyphenyl)ethylamine
Formel:
C₁₀H₁₅NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06761897 CAS-Nummer: 100570-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...