Methyl isoquinoline-5-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-Isoquinolinecarboxylate
OR925357-1G
APOSOR925357-1GEA
60.9
EUR
APOSOR925357-1G
Methyl isoquinoline-5-carboxylate 95%
Methyl isoquinoline-5-carboxylate
Formel:
C₁₁H₉NO₂ Molecular Weight: 187.195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204278 CAS-Nummer: 16675-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...