2-Methoxy-N-methylbenzylamine 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Methoxyphenyl)-N-methylmethanamine

OR925436-5G
APOSOR925436-5GEA 89.9 EUR
APOSOR925436-5G
2-Methoxy-N-methylbenzylamine 95%
2-Methoxy-N-methylbenzylamine
Formel: C₉H₁₃NO
Molecular Weight: 151,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025755
CAS-Nummer: 6851-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR