4-Hydroxybenzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

OR925449-1G OR925449-5G
APOSOR925449-1GEA 0 EUR
APOSOR925449-1G APOSOR925449-5G
4-Hydroxybenzenesulphonyl chloride 97%
4-Hydroxybenzenesulphonyl chloride
Formel: C₆H₅ClO₃S
Lagertemperatur: Kühlschrank
MDL: MFCD09833838
CAS-Nummer: 4025-67-6
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR