(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-Isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, (2R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine

OR925455-5G
APOSOR925455-5GEA 195 EUR
APOSOR925455-5G
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazin 97%
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazin
Formel: C₉H₁₆N₂O₂
Molecular Weight: 184.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040565
CAS-Nummer: 109838-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR