6-Acetyl-2-Benzoxazolinone 95%

Lieferant: Apollo Scientific

Synonyme: 6-Acetyl-2-benzoxazolinone, 6-Acetyl-2(3H)-benzoxazolone, 6-Acetyl-2-hydroxybenzoxazole, 6-Acetyl-2-benzoxazolone, 6-Acetylbenzo[d]oxazol-2(3H)-one

OR925460-5G
APOSOR925460-5GEA 157 EUR
APOSOR925460-5G
6-Acetyl-2-Benzoxazolinone 95%
6-Acetyl-2-Benzoxazolinone
Formel: C₉H₇NO₃
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01664312
CAS-Nummer: 54903-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR