3'-Hydroxypropiophenone 97%

Lieferant: Apollo Scientific

Synonyme: 1-(3-Hydroxyphenyl)propan-1-one

OR925473-100G
APOSOR925473-100GEA 132 EUR
APOSOR925473-100G
3'-Hydroxypropiophenone 97%
3'-Hydroxypropiophenone
Formel: C₉H₁₀O₂
Molecular Weight: 150.175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428514
CAS-Nummer: 13103-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR