(1-Methylnaphthalen-4-yl)methanol 95%

Lieferant: Apollo Scientific

Synonyme: (4-Methylnaphthalen-1-yl)methanol

OR925505-1G
APOSOR925505-1GEA 30.6 EUR
APOSOR925505-1G
(1-Methylnaphthalen-4-yl)methanol 95%
(1-Methylnaphthalen-4-yl)methanol
Formel: C₁₂H₁₂O
Molecular Weight: 172.223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00155252
CAS-Nummer: 57322-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR