Ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate 95%

Lieferant: Apollo Scientific

OR925561-1G
APOSOR925561-1GEA 632 EUR
APOSOR925561-1G
Ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate
Formel: C₇H₉ClN₂O₂
Molecular Weight: 188,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07357430
CAS-Nummer: 137487-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR